ENA429361422 Enamine

(2R,3R)-1-cyclopropyl-2-(3,4-difluorophenyl)-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD28657111
  • Cas Number : 1820579-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192991
Availability details for
Quantity
  • Availability
  • Add to Cart