ENA429361425 Enamine

2-(decylsulfanyl)-4,5-dihydro-1H-imidazole hydrobromide

  • MDL number MFCD28125304
  • Cas Number : 1803582-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61987
Availability details for
Quantity
  • Availability
  • Add to Cart