ENA429361427 Enamine

3-chloro-4-(2,3-dichlorophenoxy)benzoic acid

  • MDL number MFCD18584793
  • Cas Number : 1406477-36-8
  • Molecular Weight : 317.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-192595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192595
Availability details for
Quantity
  • Availability
  • Add to Cart