ENA429361428 Enamine

6-(trifluoromethyl)-5,6,7,8-tetrahydroquinazolin-2-amine

  • MDL number MFCD20410282
  • Cas Number : 1342853-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192440
Availability details for
Quantity
  • Availability
  • Add to Cart