ENA429361429 Enamine

N-(2-aminoethyl)-4-iodobenzamide hydrochloride

  • MDL number MFCD28125259
  • Cas Number : 145343-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192597
Availability details for
Quantity
  • Availability
  • Add to Cart