ENA429361430 Enamine

3-(oxan-3-yl)-3-oxopropanenitrile

  • MDL number MFCD17275101
  • Cas Number : 1343316-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192598
Availability details for
Quantity
  • Availability
  • Add to Cart