ENA429361432 Enamine

2-(aminomethyl)-4-methylpentanoic acid hydrochloride

  • MDL number MFCD28125260
  • Cas Number : 685898-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192600
Availability details for
Quantity
  • Availability
  • Add to Cart