ENA429361433 Enamine

methyl 2-(2-aminoacetamido)acetate hydrochloride

  • MDL number MFCD00039056
  • Cas Number : 2776-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192601

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-192601
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart