ENA429361434 Enamine

2-[4-(2-oxo-1,3-oxazolidin-3-yl)phenyl]acetic acid

  • MDL number MFCD11167845
  • Cas Number : 92288-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192602
Availability details for
Quantity
  • Availability
  • Add to Cart