ENA429361436 Enamine

4-(2-fluorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD14690896
  • Cas Number : 1248955-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192445
Availability details for
Quantity
  • Availability
  • Add to Cart