ENA429361440 Enamine

1-(2,3-dihydro-1-benzofuran-2-carbonyl)piperazine

  • MDL number MFCD09806692
  • Cas Number : 1016733-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192649
Availability details for
Quantity
  • Availability
  • Add to Cart