ENA429361448 Enamine

1-(6-amino-2,3-dimethoxyphenyl)ethan-1-one

  • MDL number MFCD20690878
  • Cas Number : 98300-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192410
Availability details for
Quantity
  • Availability
  • Add to Cart