ENA429361453 Enamine

1-(2,2,5,5-tetramethyloxolan-3-yl)-1H-pyrazole

  • MDL number MFCD28125265
  • Cas Number : 1601130-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192730
Availability details for
Quantity
  • Availability
  • Add to Cart