ENA429361455 Enamine

1-(bromomethyl)-4-chloro-2-methoxybenzene

  • MDL number MFCD09744685
  • Cas Number : 76283-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192732

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-192732
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart