ENA429361457 Enamine

2-(4-fluorophenyl)-4,6-dimethylpyrimidine

  • MDL number MFCD11218619
  • Cas Number : 1188093-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192734
Availability details for
Quantity
  • Availability
  • Add to Cart