ENA429361464 Enamine

(1,4-dimethylcyclohexyl)methanamine

  • MDL number MFCD21197334
  • Cas Number : 1511900-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192750
Availability details for
Quantity
  • Availability
  • Add to Cart