ENA429361465 Enamine

N4-butyl-6-chloropyrimidine-2,4-diamine

  • MDL number MFCD15201686
  • Cas Number : 5457-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192468
Availability details for
Quantity
  • Availability
  • Add to Cart