ENA429361466 Enamine

6-methylpyridine-2-carbohydrazide

  • MDL number MFCD09736941
  • Cas Number : 98547-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192757

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192757
Availability details for
Quantity
  • Availability
  • Add to Cart