ENA429361467 Enamine

5-chloro-3,4-dihydro-1H-2-benzopyran-4-one

  • MDL number MFCD09744066
  • Cas Number : 1260859-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192758
Availability details for
Quantity
  • Availability
  • Add to Cart