ENA429361471 Enamine

N-(4-cyclopropyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD01794602
  • Cas Number : 324579-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192768
Availability details for
Quantity
  • Availability
  • Add to Cart