ENA429361478 Enamine

1-(3-aminophenyl)-4-methyl-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD24108705
  • Cas Number : 1519859-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192792
Availability details for
Quantity
  • Availability
  • Add to Cart