ENA429361479 Enamine

3-(pyrrolidine-1-carbonyl)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD10661945
  • Cas Number : 69454-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192416
Availability details for
Quantity
  • Availability
  • Add to Cart