ENA429361485 Enamine

8-bromo-4-chloro-7-methylquinoline-3-carbonitrile

  • MDL number MFCD24150694
  • Cas Number : 1526860-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192811
Availability details for
Quantity
  • Availability
  • Add to Cart