ENA429361488 Enamine

4-(3,5-dimethoxyphenyl)piperidine hydrochloride

  • MDL number MFCD08751543
  • Cas Number : 1198285-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192428

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192428
Availability details for
Quantity
  • Availability
  • Add to Cart