ENA429361494 Enamine

4-bromo-2-chloro-5-fluorobenzoic acid

  • MDL number MFCD00236589
  • Cas Number : 177480-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192856
Availability details for
Quantity
  • Availability
  • Add to Cart