ENA429361495 Enamine

methyl 4-bromo-2-chloro-5-fluorobenzoate

  • MDL number MFCD12913461
  • Cas Number : 908248-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192857
Availability details for
Quantity
  • Availability
  • Add to Cart