ENA429361497 Enamine

methyl 2-amino-4-bromo-5-fluorobenzoate

  • MDL number MFCD22565660
  • Cas Number : 1395493-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192859
Availability details for
Quantity
  • Availability
  • Add to Cart