ENA429361498 Enamine

5-acetyl-2,4-dimethylbenzene-1-sulfonamide

  • MDL number MFCD18897459
  • Cas Number : 1339526-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192873
Availability details for
Quantity
  • Availability
  • Add to Cart