ENA429361499 Enamine

(4-fluoro-2-methylphenyl)thiourea

  • MDL number MFCD00041181
  • Cas Number : 946612-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192511
Availability details for
Quantity
  • Availability
  • Add to Cart