ENA429361501 Enamine

2-(4,5-dibromofuran-2-yl)pyrimidin-5-amine

  • MDL number MFCD16672196
  • Cas Number : 1249123-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192878
Availability details for
Quantity
  • Availability
  • Add to Cart