ENA429361504 Enamine

2-({bicyclo[2.2.1]heptan-2-yl}amino)ethan-1-ol

  • MDL number MFCD14622578
  • Cas Number : 37031-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192514
Availability details for
Quantity
  • Availability
  • Add to Cart