ENA429361506 Enamine

N1-(pyridin-4-yl)benzene-1,2-diamine

  • MDL number MFCD02178028
  • Cas Number : 65053-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192515
Availability details for
Quantity
  • Availability
  • Add to Cart