ENA429361512 Enamine

(5-fluoro-6-oxo-1,6-dihydropyridin-3-yl)boronic acid

  • MDL number MFCD12964544
  • Cas Number : 1141886-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192393
Availability details for
Quantity
  • Availability
  • Add to Cart