ENA429361515 Enamine

(3R,6R)-6-methylpiperidin-3-ol

  • MDL number MFCD19238007
  • Cas Number : 285995-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192536

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192536
Availability details for
Quantity
  • Availability
  • Add to Cart