ENA429361520 Enamine

(3-bromo-2-methylphenyl)boronic acid

  • MDL number MFCD18447597
  • Cas Number : 1184298-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192558
Availability details for
Quantity
  • Availability
  • Add to Cart