ENA429361521 Enamine

5-fluoro-1-(propan-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD18814482
  • Cas Number : 146366-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192559
Availability details for
Quantity
  • Availability
  • Add to Cart