ENA429361538 Enamine

1-chloro-2-methanesulfonylethane

  • MDL number MFCD11934323
  • Cas Number : 50890-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 9-Aug
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-22409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22409
Availability details for
Quantity
  • Availability
  • Add to Cart