ENA429361545 Enamine

cyclopent-1-ene-1-carbonitrile

  • MDL number MFCD00043765
  • Cas Number : 3047-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30399

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30399
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart