ENA429361549 Enamine

ethyl 2-amino-2-(4-chlorophenyl)acetate hydrochloride

  • MDL number MFCD09289493
  • Cas Number : 80589-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34572

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-34572
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart