ENA429361553 Enamine

ethyl 2-(chlorosulfonyl)benzoate

  • MDL number MFCD01961366
  • Cas Number : 103008-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35954
Availability details for
Quantity
  • Availability
  • Add to Cart