ENA429361557 Enamine

(naphthalen-2-yl)methanamine hydrochloride

  • MDL number MFCD00137202
  • Cas Number : 2241-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 266-270
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39736
Availability details for
Quantity
  • Availability
  • Add to Cart