ENA429361560 Enamine

1-(3-aminophenyl)-3-propylurea

  • MDL number MFCD11171878
  • Cas Number : 197644-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41694
Availability details for
Quantity
  • Availability
  • Add to Cart