ENA429361571 Enamine

N-benzyl-N-methylacetamide

  • MDL number MFCD01195308
  • Cas Number : 29823-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49558
Availability details for
Quantity
  • Availability
  • Add to Cart