ENA429361580 Enamine

1-(3-chloropyridin-2-yl)-4-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD28118881
  • Cas Number : 1708455-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55671
Availability details for
Quantity
  • Availability
  • Add to Cart