ENA429361586 Enamine

3,8-dimethyl-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD16663807
  • Cas Number : 1174645-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57777
Availability details for
Quantity
  • Availability
  • Add to Cart