ENA429361590 Enamine

4-(piperidin-3-yl)pyrimidin-2-amine

  • MDL number MFCD16093637
  • Cas Number : 1250024-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59369
Availability details for
Quantity
  • Availability
  • Add to Cart