ENA429361591 Enamine

4-(1-bromoethyl)pyridine

  • MDL number MFCD11103123
  • Cas Number : 754218-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60221

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60221
Availability details for
Quantity
  • Availability
  • Add to Cart