ENA429361593 Enamine

3-cyclohexyl-3-methylbutanoic acid

  • MDL number MFCD20650734
  • Cas Number : 4094-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192439
Availability details for
Quantity
  • Availability
  • Add to Cart