ENA429361595 Enamine

2-(1-ethyl-1H-imidazol-5-yl)piperidine

  • MDL number MFCD28118882
  • Cas Number : 1601232-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62893
Availability details for
Quantity
  • Availability
  • Add to Cart