ENA429361606 Enamine

3-(3-cyanophenyl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD10694133
  • Cas Number : 1152531-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68396

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68396
Availability details for
Quantity
  • Availability
  • Add to Cart